Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP23 Monoclonal Antibody (OTI3C10), TrueMAB™, OriGene
SDP

Catalog No. 502540616
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0616 30 μL
50-167-4720 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0616 Supplier OriGene Supplier No. TA503944S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DUSP23 Monoclonal antibody specifically detects DUSP23 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen DUSP23
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3C10
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SDS
Gene Accession No. P20132
Gene Alias 1300005N15Rik; dual specificity phosphatase 23; dual specificity protein phosphatase 23; Dusp23; DUSP25; LDP3; LDP-3; Low molecular mass dual specificity phosphatase 3; low-molecular-mass dual-specificity phosphatase; low-molecular-mass dual-specificity phosphatase 3; MOSP; RP11-190A12.1; testicular tissue protein Li 59; VH1-like member Z; VH1-like phosphatase Z; VHZ
Host Species Mouse
Immunogen Full length human recombinant protein of human SDS produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10993
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.