Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP23 Mouse anti-Human, Clone: OTI3C10, lyophilized, TrueMAB™
SDP

Catalog No. 501662503
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-2503 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-2503 Supplier OriGene Supplier No. CF503944
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Myelin / Oligodendrocyte Specific Protein (MOSP) (pI of ∽6.7) is found exclusively on oligodendrocytes and myelin in the central nervous system (CNS). Myelin is produced by oligodendrocytes in the CNS and Schwann cells in the peripheral nervous system (PNS). Defining the differences between CNS and PNS myelin may provide insights into the process of myelination and the pathogenesis of certain idiopathic diseases involving myelin such as multiple sclerosis, certain leukodystrophies, and other myelinopathies. On demyelination, immunoreactivity for MOSP, like MBP, is lost.

Specifications

Antigen DUSP23
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3C10
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene DUSP23
Gene Accession No. Q9BVJ7
Gene Alias DUSP25, LDP-3, MOSP, VHZ
Gene Symbols DUSP23
Host Species Mouse
Immunogen Full length human recombit protein of human DUSP23 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54935
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.