Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DUSP23 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533490
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%) Marmoset, Horse, Rabbit (94%) Elephant, Bat (89%) Rat, Hamster, Bovine (83%).
Protein phosphatase that mediates dephosphorylation of proteins phosphorylated on Tyr and Ser/Thr residues. In vitro, it can dephosphorylate p44-ERK1 (MAPK3) but not p54 SAPK-beta (MAPK10) in vitro. Able to enhance activation of JNK and p38 (MAPK14).
Specifications
| DUSP23 | |
| Polyclonal | |
| Unconjugated | |
| DUSP23 | |
| 1300005N15Rik; dual specificity phosphatase 23; dual specificity protein phosphatase 23; Dusp23; DUSP25; LDP3; LDP-3; Low molecular mass dual specificity phosphatase 3; low-molecular-mass dual-specificity phosphatase; low-molecular-mass dual-specificity phosphatase 3; MOSP; RP11-190A12.1; testicular tissue protein Li 59; VH1-like member Z; VH1-like phosphatase Z; VHZ | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54935 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9BVJ7 | |
| DUSP23 | |
| Synthetic 18 amino acid peptide from internal region of human DUSP23. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction