Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUSP23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583793
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583793 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583793 Supplier Invitrogen™ Supplier No. PA583793
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IVDEANARGE AVGVHCALGF GRTGTMLACY LVKERGLAAG DAIAEIRRLR PGSIETYEQE KAVFQFYQRT K Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 97%.

Protein phosphatase that mediates dephosphorylation of proteins phosphorylated on Tyr and Ser/Thr residues. In vitro, it can dephosphorylate p44-ERK1 (MAPK3) but not p54 SAPK-beta (MAPK10) in vitro. Able to enhance activation of JNK and p38 (MAPK14).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP23
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DUSP23
Gene Accession No. Q9BVJ7
Gene Alias 1300005N15Rik; dual specificity phosphatase 23; dual specificity protein phosphatase 23; Dusp23; DUSP25; LDP3; LDP-3; Low molecular mass dual specificity phosphatase 3; low-molecular-mass dual-specificity phosphatase; low-molecular-mass dual-specificity phosphatase 3; MOSP; RP11-190A12.1; testicular tissue protein Li 59; VH1-like member Z; VH1-like phosphatase Z; VHZ
Gene Symbols DUSP23
Host Species Rabbit
Immunogen Recombinant protein fragment of Human DUSP23 (Product #RP-94328)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54935
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.