Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP26 Mouse anti-Human, Polyclonal, Abnova™

Catalog No. 89335049 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-049 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-049 Supplier Abnova Corporation Supplier No. H00078986B01P
Add to Cart
Add to Cart

Mouse Polyclonal Antibody.

DUSP26 Polyclonal antibody specifically detects DUSP26 in Human samples. It is validated for WB-Tr.

Specifications

Antigen DUSP26
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description dual specificity phosphatase 26 (putative)
Gene DUSP26
Gene Accession No. NM_024025.1
Gene Alias DUSP24, LDP-4, MGC1136, MGC2627, MKP8, NATA1, SKRP3
Host Species Mouse
Immunogen DUSP26 (NP_076930.1, 1 a.a. ∼ 211 a.a) full-length human protein
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 78986
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.