Learn More
Invitrogen™ DUSP26 Polyclonal Antibody

Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | DUSP26 |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.52 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | DUSP26 |
| Gene Accession No. | Q5FVI9, Q9BV47, Q9D700 |
| Gene Alias | 2310043K02Rik; DSP-4; dual specificity phosphatase 26 (putative); dual specificity phosphatase SKRP3; Dual specificity protein phosphatase 26; dual-specificity phosphatase SKRP3; DUSP24; DUSP26; LDP4; LDP-4; Low-molecular-mass dual-specificity phosphatase 4; MAP kinase phosphatase 8; MGC1136; MGC2627; Mitogen-activated protein kinase phosphatase 8; MKP8; MKP-8; NATA1; NEAP; neuroendocrine-associated phosphatase; Novel amplified gene in thyroid anaplastic cancer; RGD1310090; Skrp3 |
| Gene Symbols | DUSP26 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.