Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUSP26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532739
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532739 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532739 Supplier Invitrogen™ Supplier No. PA532739
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Marmoset, Mouse, Rat, Bovine, Hamster, Elephant, Panda, Horse, Rabbit, Opossum (100%); Bat, Pig (94%); Platypus (89%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

This gene encodes a member of the tyrosine phosphatase family of proteins and exhibits dual specificity by dephosphorylating tyrosine as well as serine and threonine residues. This gene has been described as both a tumor suppressor and an oncogene depending on the cellular context. This protein may regulate neuronal proliferation and has been implicated in the progression of glioblastoma through its ability to dephosphorylate the p53 tumor suppressor. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Feb 2015].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP26
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene DUSP26
Gene Accession No. Q17QJ3, Q5FVI9, Q9BV47, Q9D700
Gene Alias 2310043K02Rik; DSP-4; dual specificity phosphatase 26 (putative); dual specificity phosphatase SKRP3; Dual specificity protein phosphatase 26; dual-specificity phosphatase SKRP3; DUSP24; DUSP26; LDP4; LDP-4; Low-molecular-mass dual-specificity phosphatase 4; MAP kinase phosphatase 8; MGC1136; MGC2627; Mitogen-activated protein kinase phosphatase 8; MKP8; MKP-8; NATA1; NEAP; neuroendocrine-associated phosphatase; Novel amplified gene in thyroid anaplastic cancer; RGD1310090; Skrp3
Gene Symbols DUSP26
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from internal region of human DUSP26.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100058104, 100340562, 100761425, 306527, 533896, 66959, 78986
Target Species Human, Mouse, Rat, Rabbit, Monkey, Bovine, Hamster, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.