Learn More
Invitrogen™ DUSP26 Polyclonal Antibody

Description
Immunogen sequence: CPGNWLWASM TFMARFSRSS SRSPVRTRGT LEEMPTVQHP FLNVFELERL LYTGKTACNH ADEVWPGLYL GDQDMANNRR ELRRLGITHV L Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 97%.
Specifications
Specifications
| Antigen | DUSP26 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | DUSP26 |
| Gene Accession No. | Q9BV47 |
| Gene Alias | 2310043K02Rik; DSP-4; dual specificity phosphatase 26 (putative); dual specificity phosphatase SKRP3; Dual specificity protein phosphatase 26; dual-specificity phosphatase SKRP3; DUSP24; DUSP26; LDP4; LDP-4; Low-molecular-mass dual-specificity phosphatase 4; MAP kinase phosphatase 8; MGC1136; MGC2627; Mitogen-activated protein kinase phosphatase 8; MKP8; MKP-8; NATA1; NEAP; neuroendocrine-associated phosphatase; Novel amplified gene in thyroid anaplastic cancer; RGD1310090; Skrp3 |
| Gene Symbols | DUSP26 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.