Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP26 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA132629
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA132629 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA132629 Supplier Thermo Scientific Supplier No. PA132629

Rabbit Polyclonal Antibody

PA1-32629 detects DUSP26 from human samples.

This gene encodes a member of the tyrosine phosphatase family of proteins and exhibits dual specificity by dephosphorylating tyrosine as well as serine and threonine residues. This gene has been described as both a tumor suppressor and an oncogene depending on the cellular context. This protein may regulate neuronal proliferation and has been implicated in the progression of glioblastoma through its ability to dephosphorylate the p53 tumor suppressor. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Feb 2015].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP26
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene DUSP26
Gene Accession No. Q9BV47
Gene Alias 2310043K02Rik; DSP-4; dual specificity phosphatase 26 (putative); dual specificity phosphatase SKRP3; Dual specificity protein phosphatase 26; dual-specificity phosphatase SKRP3; DUSP24; DUSP26; LDP4; LDP-4; Low-molecular-mass dual-specificity phosphatase 4; MAP kinase phosphatase 8; MGC1136; MGC2627; Mitogen-activated protein kinase phosphatase 8; MKP8; MKP-8; NATA1; NEAP; neuroendocrine-associated phosphatase; Novel amplified gene in thyroid anaplastic cancer; RGD1310090; Skrp3
Gene Symbols DUSP26
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from near C-terminus of human DUSP26
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78986
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.