Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUSP27/DUPD1 Antibody (OTI7E4), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272503APC
Description
DUSP27/DUPD1 Monoclonal specifically detects DUSP27/DUPD1 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| DUSP27/DUPD1 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| DUPD1 | |
| Full length recombinant protein of human DUSP27/DUPD1 (NP_001003892) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Flow Cytometry | |
| OTI7E4 | |
| Flow Cytometry | |
| atypical dual-specific protein phosphatase, Dual specificity phosphatase 27, dual specificity phosphatase and pro isomerase domain containing 1, DUSP27dual specificity phosphatase DUPD1, EC 3.1.3.16, EC 3.1.3.48, FMDSP | |
| Mouse | |
| Affinity Purified | |
| Protein Phosphatase | |
| 338599.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction