Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUSP27/DUPD1 Antibody (OTI7E4), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272503MFV610
Description
DUSP27/DUPD1 Monoclonal antibody specifically detects DUSP27/DUPD1 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
DUSP27/DUPD1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
OTI7E4 | |
Flow Cytometry | |
atypical dual-specific protein phosphatase, Dual specificity phosphatase 27, dual specificity phosphatase and pro isomerase domain containing 1, DUSP27dual specificity phosphatase DUPD1, EC 3.1.3.16, EC 3.1.3.48, FMDSP | |
Full length human recombinant protein of human DUPD1(NP_001003892) produced in HEK293T cell. | |
0.1 mL | |
Protein Phosphatase | |
338599 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction