Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUPD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560176
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560176 Supplier Invitrogen™ Supplier No. PA560176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTSGEVKTSL KNAYSSAKRL SPKMEEEGEE EDYCTPGAFE LERLFWKGSP QYTHVNEVWP KLYIGDEATA LDRYRL Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 76%.

Dual specificity phosphatase DUPD1 is able to dephosphorylate phosphotyrosine, phosphoserine and phosphothreonine residues, with a preference for phosphotyrosine as a substrate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUPD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DUPD1
Gene Accession No. Q68J44
Gene Alias atypical dual-specific protein phosphatase; dual specificity phosphatase 27; dual specificity phosphatase and pro isomerase domain containing 1; dual specificity phosphatase DUPD1; DUPD1; DUSP27; FMDSP
Gene Symbols DUPD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DUPD1. Recombinant protein control fragment (Product #RP-91585).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338599
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.