Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DUSP27 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564945
Description
Immunogen sequence: EALMTVRKKRA IYPNEGFLKQ LRELNEKLME ER Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
DUSP27 (Dual Specificity Phosphatase 27, Atypical) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include phosphatase activity and protein tyrosine/serine/threonine phosphatase activity. An important paralog of this gene is DUSP13.
Specifications
DUSP27 | |
Polyclonal | |
Unconjugated | |
DUSP27 | |
atypical dual-specific protein phosphatase; dual specificity phosphatase 27; dual specificity phosphatase 27 (putative); dual specificity phosphatase and pro isomerase domain containing 1; dual specificity phosphatase DUPD1; DUPD1; DUSP27; FMDSP; Inactive dual specificity phosphatase 27 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
92235 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VZP5 | |
DUSP27 | |
Recombinant Human DUSP27. Recombinant protein control fragment (Product #RP-90298). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction