Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP28 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA561523
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561523 Supplier Thermo Scientific Supplier No. PA561523

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Immunogen sequence: GACLVYCKNG RSRSAAVCTA YLMRHRGLSL AKAFQMVKSA RPVAEPNPGF WSQLQKYEEA LQAQSCLQGE PPALGLGPEA Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

DUSP28 Polyclonal antibody specifically detects DUSP28 in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP28
Applications Immunohistochemistry
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DUSP28
Gene Accession No. Q4G0W2
Gene Alias Dual specificity phosphatase 28; DUSP 28; DUSP26; DUSP28; DUSP-28; VHP
Gene Symbols DUSP28
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DUSP28
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285193
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.