Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUSP5 Monoclonal Antibody (OTI5H9)
GREENER_CHOICE

Catalog No. PIMA527383
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527383 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527383 Supplier Invitrogen™ Supplier No. MA527383
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DUSP5 Monoclonal Antibody for Western Blot

The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein kinase superfamily, which are associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product inactivates ERK1, is expressed in a variety of tissues with the highest levels in pancreas and brain, and is localized in the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP5
Applications Western Blot
Classification Monoclonal
Clone OTI5H9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DUSP5
Gene Accession No. O54838, Q16690
Gene Alias candidate plasticity-related gene 21; Cpg21; dual specificity phosphatase 5; dual specificity protein phosphatase 5; dual specificity protein phosphatase 5-like; dual specificity protein phosphatase hVH3; DUSP; DUSP5; Gm337; HVH3; LOW QUALITY PROTEIN: dual specificity protein phosphatase 5; MAP-kinase phosphatase (cpg21); MAP-kinase phosphatase CPG21; serine/threonine specific protein phosphatase; VH1-like phosphatase 3; VH3
Gene Symbols DUSP5
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 125-384 of DUSP5 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171109, 1847
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.