Learn More
DUSP5 Mouse anti-Human, Clone: OTI2F8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811034

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein kinase superfamily, which are associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product inactivates ERK1, is expressed in a variety of tissues with the highest levels in pancreas and brain, and is localized in the nucleus.Specifications
DUSP5 | |
Monoclonal | |
Unconjugated | |
DUSP5 | |
DUSP, HVH3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1847 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2F8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q16690 | |
DUSP5 | |
Human recombit protein fragment corresponding to amino acids 125-384 of human DUSP5 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.