Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100490
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100490 Supplier Thermo Scientific Supplier No. PA5100490
Add to Cart
Add to Cart

DUSP7 Polyclonal Antibody

DUSP7 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DUSP7
Gene Accession No. Q16829, Q63340, Q91Z46
Gene Alias AU015694; dual specificity phosphatase 6; dual specificity phosphatase 7; dual specificity protein phosphatase 7; Dual specificity protein phosphatase MKP-X; dual specificity protein phosphatase PYST2; dual-specificity phosphatase-7; DUSP7; fd19d04; hypothetical protein LOC327246; MAP kinase phosphatase; Mkpx; mkp-X; PYST2; wu:fd19d04; zgc:55957; zgc:77211; zgc:77211 protein
Gene Symbols DUSP7
Host Species Rabbit
Immunogen A synthesized peptide derived from human DUSP7(Accession Q16829), corresponding to amino acid residues S197-Q247.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1849, 235584, 300980
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.