Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUSP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109668
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109668 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109668 Supplier Invitrogen™ Supplier No. PA5109668
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NleAGDRLPR KVNl.

The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which is associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product inactivates SAPK/JNK and p38, is expressed predominantly in the adult brain, heart, and skeletal muscle, is localized in the cytoplasm, and is induced by nerve growth factor and insulin. An intronless pseudogene for DUSP8 is present on chromosome 10q11.2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP8
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene DUSP8
Gene Accession No. O09112, Q13202
Gene Alias 5530400B01Rik; AI593498; C11orf81; dual specificity phosphatase 8; dual specificity protein phosphatase 8; Dual specificity protein phosphatase hVH-5; Dusp8; FLJ42476; FLJ42958; H1 phosphatase, vaccinia virus homolog; Hb5; HVH-5; HVH8; M3/6; neuronal tyrosine threonine phosphatase 1; neuronal tyrosine/threonine phosphatase 1; Nttp1; OTTHUMP00000164487; serine/threonine specific protein phosphatase; VH5
Gene Symbols DUSP8
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18218, 1850, 361679
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.