Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUT Recombinant Rabbit Monoclonal Antibody (HL1210)
GREENER_CHOICE

Catalog No. PIMA546959
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546959 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA546959 Supplier Invitrogen™ Supplier No. MA546959
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

This gene encodes an essential enzyme of nucleotide metabolism. The encoded protein forms a ubiquitous, homotetrameric enzyme that hydrolyzes dUTP to dUMP and pyrophosphate. This reaction serves two cellular purposes: providing a precursor (dUMP) for the synthesis of thymine nucleotides needed for DNA replication, and limiting intracellular pools of dUTP. Elevated levels of dUTP lead to increased incorporation of uracil into DNA, which induces extensive excision repair mediated by uracil glycosylase. This repair process, resulting in the removal and reincorporation of dUTP, is self-defeating and leads to DNA fragmentation and cell death. Alternative splicing of this gene leads to different isoforms that localize to either the mitochondrion or nucleus. A related pseudogene is located on chromosome 19.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1210
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Dut
Gene Accession No. P33316
Gene Alias 5031412I06Rik; 5133400F09Rik; D2Bwg0749e; Deoxyuridine 5'-triphosphate nucleotidohydrolase; deoxyuridine 5'-triphosphate nucleotidohydrolase, mitochondrial; deoxyuridine triphosphatase; Dut; Dutp; dUTP diphosphatase; dUTP nucleotidohydrolase; dUTP pyrophosphatase; dUTPase; FLJ20622; PIP4; PPAR-interacting protein 4
Gene Symbols Dut
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DUT. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1854
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.