Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA129808
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA129808 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA129808 Supplier Invitrogen™ Supplier No. PA129808
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-29808 detects DUT-N from human samples.

This gene encodes an essential enzyme of nucleotide metabolism. The encoded protein forms a ubiquitous, homotetrameric enzyme that hydrolyzes dUTP to dUMP and pyrophosphate. This reaction serves two cellular purposes: providing a precursor (dUMP) for the synthesis of thymine nucleotides needed for DNA replication, and limiting intracellular pools of dUTP. Elevated levels of dUTP lead to increased incorporation of uracil into DNA, which induces extensive excision repair mediated by uracil glycosylase. This repair process, resulting in the removal and reincorporation of dUTP, is self-defeating and leads to DNA fragmentation and cell death. Alternative splicing of this gene leads to different isoforms that localize to either the mitochondrion or nucleus. A related pseudogene is located on chromosome 19.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUT
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.02% sodium azide
Gene Dut
Gene Accession No. P33316
Gene Alias 5031412I06Rik; 5133400F09Rik; D2Bwg0749e; Deoxyuridine 5'-triphosphate nucleotidohydrolase; deoxyuridine 5'-triphosphate nucleotidohydrolase, mitochondrial; deoxyuridine triphosphatase; Dut; Dutp; dUTP diphosphatase; dUTP nucleotidohydrolase; dUTP pyrophosphatase; dUTPase; FLJ20622; PIP4; PPAR-interacting protein 4
Gene Symbols Dut
Host Species Rabbit
Immunogen Mixture of KLH-conjugated synthetic peptides corresponding to amino acids 25-44 and 113-130 of human dUTPase.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1854
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.