Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dUTPase Antibody (SR1280), Novus Biologicals™
SDP

Catalog No. NB260456 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260456 100 μg
NB260027 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB260456 Supplier Novus Biologicals Supplier No. NBP322634100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

dUTPase Monoclonal antibody specifically detects dUTPase in Human samples. It is validated for Western Blot, Immunofluorescence, Serum

Specifications

Antigen dUTPase
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone SR1280
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence, Serum 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias deoxyuridine triphosphatase, dUTP pyrophosphatasedUTP nucleotidohydrolase, dUTPasedeoxyuridine 5'-triphosphate nucleotidohydrolase, mitochondrial, EC 3.6.1.23, FLJ20622
Host Species Rabbit
Immunogen A synthesized peptide derived from human dUTPase (Uniprot #: P33316)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1854
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.