Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUX4 Monoclonal Antibody (P2B1), FITC
GREENER_CHOICE

Catalog No. PIMA545266
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545266 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545266 Supplier Invitrogen™ Supplier No. MA545266
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45266 was sufficient for detection of DUX4 in 20 μg of HeLa cell lysate by ECL immunoblot analysis using goat anti-mouse IgG: HRP as teh secondary. Detects approximately 45kDa. No cross-reactivity with DUX4c.

DUX4 (also known as Double homeobox 4) is the leading candidate causative gene for facioscapulohumeral dystrophy (FSHD), a degenerative skeletal muscle disease and one of the most common muscular dystrophies. FSHD is caused by the deletion of a subset of D4Z4 macrosatellite repeats on chromosome 4. Each repeat contains a retrogene encoding the double-homeobox factor DUX4. DUX4 expression is epigenetically suppressed in differentiated tissues and the residual DUX4 transcripts are spliced to remove the carboxyterminal domain that has been associated with cell toxicity. In FSHD individuals, the expression of the full-length DUX4 transcript is not completely suppressed in skeletal muscle, and possibly other differentiated tissues, and results in a small percentage of cells expressing relatively abundant amounts of the full-length DUX4 mRNA and protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUX4
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone P2B1
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene Dux4
Gene Accession No. Q9UBX2
Gene Alias AW822073; double homeobox; double homeobox 4; double homeobox 4-like; double homeobox B-like 1; double homeobox protein 10; Double homeobox protein 4; double homeobox protein 4/10; Dux; DUX10; Dux4; DUX4L; Duxbl; Duxbl1; EG664783; homeobox protein; RGD1311053
Gene Symbols Dux4
Host Species Mouse
Immunogen C-terminal 76 amino acids of DUX4 with glutathione-s-transferase (gst) tag.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100288687
Target Species Human, Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.