Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DUX4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566173
Description
Immunogen sequence: PSQSEALRACF ERNPYPGIAT RER Highest antigen sequence identity to the following orthologs - mouse 58%, rat 50%.
DUX4 (also known as Double homeobox 4) is the leading candidate causative gene for facioscapulohumeral dystrophy (FSHD), a degenerative skeletal muscle disease and one of the most common muscular dystrophies. FSHD is caused by the deletion of a subset of D4Z4 macrosatellite repeats on chromosome 4. Each repeat contains a retrogene encoding the double-homeobox factor DUX4. DUX4 expression is epigenetically suppressed in differentiated tissues and the residual DUX4 transcripts are spliced to remove the carboxyterminal domain that has been associated with cell toxicity. In FSHD individuals, the expression of the full-length DUX4 transcript is not completely suppressed in skeletal muscle, and possibly other differentiated tissues, and results in a small percentage of cells expressing relatively abundant amounts of the full-length DUX4 mRNA and protein.
Specifications
DUX4 | |
Polyclonal | |
Unconjugated | |
Dux4 | |
AW822073; double homeobox; double homeobox 4; double homeobox 4-like; double homeobox B-like 1; double homeobox protein 10; Double homeobox protein 4; double homeobox protein 4/10; Dux; DUX10; Dux4; DUX4L; Duxbl; Duxbl1; EG664783; homeobox protein; RGD1311053 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100288687 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UBX2 | |
Dux4 | |
Recombinant Human DUX4. Recombinant protein control fragment (Product #RP-106857). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction