Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUX4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310390100UL
Description
DUX4 Polyclonal specifically detects DUX4 in Human samples. It is validated for Western Blot.Specifications
DUX4 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
double homeobox 4, double homeobox protein 10, double homeobox protein 4, double homeobox protein 4/10, double homeobox protein DUX10, DUX10 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of human DUX4 (NP_001280727.1). Peptide sequence TRGAEGTLGGRQGGREAGRNGGRDRATQGLGAGPREPGTDGGRKAGRKSG | |
100 μg | |
Neuroscience, Transcription Factors and Regulators | |
100288687 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction