Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUXA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543054
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543054 Supplier Invitrogen™ Supplier No. PA543054
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SRLLLQRKRE PVASLEQEEQ GKIPEGLQGA EDTQNGTNFT SDSHFSGART Sequence homology: Human: 100%.

Homeobox genes encode DNA-binding proteins, many of which are thought to be involved in early embryonic development. Homeobox genes encode a DNA-binding domain of 60 to 63 amino acids referred to as the homeodomain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUXA
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DUXA
Gene Accession No. A6NLW8
Gene Alias double homeobox A; double homeobox protein A; DUXA
Gene Symbols DUXA
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human DUXA.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 503835
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.