Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DVL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102836
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102836 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102836 Supplier Invitrogen™ Supplier No. PA5102836
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DVL1.

DVL1, the human homolog of the Drosophila dishevelled gene (dsh) encodes a cytoplasmic phosphoprotein that regulates cellproliferation, acting as a transducer molecule for developmentalprocesses, including segmentation and neuroblast specification.DVL1 is a candidate gene for neuroblastomatous transformation. TheSchwartz-Jampel syndrome and Charcot-Marie-Tooth disease type 2Ahave been mapped to the same region as DVL1. The phenotypes ofthese diseases may be consistent with defects which might beexpected from aberrant expression of a DVL gene during development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DVL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DVL1
Gene Accession No. O14640, P51141, Q9WVB9
Gene Alias dishevelled 1; dishevelled 1 (homologous to Drosophila dsh); dishevelled segment polarity protein 1; dishevelled, dsh homolog 1; dishevelled, dsh homolog 1 (Drosophila); dishevelled-1; Dishevelled-1-like; DRS2; DSH homolog 1; DSH homolog 1-like; Dvl; Dvl1; dvl-1; DVL1L1; DVL1P1; MGC54245; mKIAA4029; RP5-890O3.5; segment polarity protein dishevelled homolog DVL-1
Gene Symbols DVL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DVL1(Accession O14640), corresponding to amino acid residues C382-R432.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13542, 1855, 83721
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.