Learn More
Invitrogen™ DVL1 Polyclonal Antibody

Description
DVL1 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | DVL1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | DVL1 |
| Gene Accession No. | O14640, P51141, Q9WVB9 |
| Gene Alias | dishevelled 1; dishevelled 1 (homologous to Drosophila dsh); dishevelled segment polarity protein 1; dishevelled, dsh homolog 1; dishevelled, dsh homolog 1 (Drosophila); dishevelled-1; Dishevelled-1-like; DRS2; DSH homolog 1; DSH homolog 1-like; Dvl; Dvl1; dvl-1; DVL1L1; DVL1P1; MGC54245; mKIAA4029; RP5-890O3.5; segment polarity protein dishevelled homolog DVL-1 |
| Gene Symbols | DVL1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.