Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DVL1P1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549516

Catalog No. PA549516


Only null left
Add to Cart

Description

Description

DVL1P1 Polyclonal Antibody for Western Blot

May play a role in the signal transduction pathway mediated by multiple Wnt genes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DVL1P1
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
P54792
DVL1P1
KLH conjugated synthetic peptide between 39-71 amino acids from human DVL1L1.
200 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
DVL1P1
Putative segment polarity protein dishevelled homolog DVL1P1, DSH homolog 1-like, Segment polarity protein dishevelled homolog DVL-1-like, Dishevelled-1-like, DVL1P1, DVL, DVL1, DVL1L1
Rabbit
Antigen affinity chromatography, Protein A
RUO
8215
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.