Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DVL1P1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597679
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597679 Supplier Invitrogen™ Supplier No. PA597679
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DVL1P1 Polyclonal Antibody for Western Blot, ELISA

May play a role in the signal transduction pathway mediated by multiple Wnt genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DVL1P1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DVL1P1
Gene Accession No. P54792
Gene Symbols DVL1P1
Host Species Rabbit
Immunogen Peptide sequence from Human Putative segment polarity protein dishevelled homolog DVL1P1 protein (152-173AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8215
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.