Learn More
Invitrogen™ DVL2 Monoclonal Antibody (OTI2E11)

Description
DVL2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | DVL2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI2E11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DVL2 |
| Gene Accession No. | O14641 |
| Gene Alias | dishevelled 2; dishevelled 2 (homologous to Drosophila dsh); dishevelled 2, dsh homolog; dishevelled 2, dsh homolog (Drosophila); dishevelled segment polarity protein 2; dishevelled, dsh homolog 2; dishevelled-2; DSH homolog 2; Dvl2; Dvl-2; LOW QUALITY PROTEIN: segment polarity protein dishevelled homolog DVL-2; sb:eu471; Segment polarity protein dishevelled homolog DVL-2; wu:fc05d12; wu:fo71e09; wu:fp54a02; zgc:55372 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.