Learn More
DVL2 Mouse anti-Human, Clone: OTI7C7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806789

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Disheveled DVL2 is a member of the dishevelled (dsh) protein family. The vertebrate dsh proteins have approximately 40% amino acid sequence similarity with Drosophila dsh. This gene encodes a 90-kD protein that undergoes posttranslational phosphorylation to form a 95-kD cytoplasmic protein, which may play a role in the signal transduction pathway mediated by multiple Wnt proteins. The mechanisms of dishevelled function in Wnt signaling are likely to be conserved among metazoans.Specifications
DVL2 | |
Monoclonal | |
Unconjugated | |
DVL2 | |
dishevelled segment polarity protein 2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1856 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7C7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O14641 | |
DVL2 | |
Human recombit protein fragment corresponding to amino acids 1-320 of human DVL2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.