Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DVL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 720060
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-006-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-006-0 Supplier Invitrogen™ Supplier No. 720060
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Rat, Mouse, Pig, Rabbit and Bovine.

Disheveled DVL2 is a member of the dishevelled (dsh) protein family. The vertebrate dsh proteins have approximately 40% amino acid sequence similarity with Drosophila dsh. This gene encodes a 90-kD protein that undergoes posttranslational phosphorylation to form a 95-kD cytoplasmic protein, which may play a role in the signal transduction pathway mediated by multiple Wnt proteins. The mechanisms of dishevelled function in Wnt signaling are likely to be conserved among metazoans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DVL2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2-7.4
Gene DVL2
Gene Accession No. O14641, Q60838
Gene Alias dishevelled 2; dishevelled 2 (homologous to Drosophila dsh); dishevelled 2, dsh homolog; dishevelled 2, dsh homolog (Drosophila); dishevelled segment polarity protein 2; dishevelled, dsh homolog 2; dishevelled-2; DSH homolog 2; Dvl2; Dvl-2; LOW QUALITY PROTEIN: segment polarity protein dishevelled homolog DVL-2; sb:eu471; Segment polarity protein dishevelled homolog DVL-2; wu:fc05d12; wu:fo71e09; wu:fp54a02; zgc:55372
Gene Symbols DVL2
Host Species Rabbit
Immunogen Peptides corresponding to Human DVL2 (aa 232-249, 596-613).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13543, 1856, 303251
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.