Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DVL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517482
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517482 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517482 Supplier Invitrogen™ Supplier No. PA517482
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with Dvl2.

This gene is a member of a multi-gene family which shares strong similarity with the Drosophila disheveled gene, dsh. The Drosophila disheveled gene encodes a cytoplasmic phosphoprotein that regulates cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DVL3
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 9 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene DVL3
Gene Accession No. Q61062, Q92997
Gene Alias dishevelled 3 (homologous to Drosophila dsh); dishevelled 3, dsh homolog; dishevelled 3, dsh homolog (Drosophila); dishevelled segment polarity protein 3; dishevelled, dsh homolog 3; dishevelled-3; DRS3; DSH homolog 3; DVL3; dvl-3; I79_017820; KIAA0208; OTTHUMP00000210778; Segment polarity protein dishevelled homolog DVL-3; Segment polarity protein dishevelled-like DVL-3
Gene Symbols DVL3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding the carboxy terminus of human Dvl3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100773535, 13544, 1857, 303811, 539203
Target Species Bovine, Hamster, Human, Mink, Mouse, Monkey, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.