Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DVL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527723
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527723 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527723 Supplier Invitrogen™ Supplier No. PA527723
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HCT116. Predicted reactivity: Mouse (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene is a member of a multi-gene family which shares strong similarity with the Drosophila disheveled gene, dsh. The Drosophila disheveled gene encodes a cytoplasmic phosphoprotein that regulates cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DVL3
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene DVL3
Gene Accession No. Q61062, Q92997
Gene Alias dishevelled 3 (homologous to Drosophila dsh); dishevelled 3, dsh homolog; dishevelled 3, dsh homolog (Drosophila); dishevelled segment polarity protein 3; dishevelled, dsh homolog 3; dishevelled-3; DRS3; DSH homolog 3; DVL3; dvl-3; I79_017820; KIAA0208; OTTHUMP00000210778; Segment polarity protein dishevelled homolog DVL-3; Segment polarity protein dishevelled-like DVL-3
Gene Symbols DVL3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human Dishevelled 3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13544, 1857
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.