Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DWARF 14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119686
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119686 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119686 Supplier Invitrogen™ Supplier No. PA5119686
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Rice. Subcellular Location: Cytoplasm, Nucleus.

Involved in strigolactone (SL) signaling pathway. May function downstream of SL synthesis, as a component of hormone signaling or as an enzyme that participates in the conversion of SL to the bioactive form. Strigolactones are hormones that inhibit tillering and shoot branching through the MAX-dependent pathway, contribute to the regulation of shoot architectural response to phosphate-limiting conditions and function as rhizosphere signal that stimulates hyphal branching of arbuscular mycorrhizal fungi and trigger seed germination of root parasitic weeds. Strigolactone-dependent association of D14 with D3 and D53 (a repressor of SL signaling) triggers D53 ubiquitination and degradation . Hydrolyzes the butenolide ring of SLs . A reaction product D-OH is trapped in the cavity of D14, inducing the interaction with SLR1, and probably with other proteins such as D3 and D53 . Contributes to the negative regulation of gibberellin signaling .
TRUSTED_SUSTAINABILITY

Specifications

Antigen DWARF 14
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias D14; Strigolactone esterase D1
Host Species Rabbit
Immunogen Synthetic peptide within N-terminal Rice D14.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.