Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWORF Rabbit anti-Mouse, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc DWORF101AP
Description
DWORF Polyclonal antibody specifically detects DWORF in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
| DWORF | |
| Polyclonal | |
| Unconjugated | |
| Strit1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 102637511 | |
| -20°C | |
| Liquid |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.5-1.5 mg/mL | |
| proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
| Sarcoplasmic/endoplasmic reticulum calcium ATPase regulator DWORF; Strit1 | |
| Synthetic peptide taken within amino acid region 1-30 on mouse Sarcoplasmic/endoplasmic reticulum calcium ATPase regulator DWORF protein. | |
| 200 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction