Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter DxH 560 Autoloader

Catalog No. 21280089 Shop All Beckman Coulter Products
Click to view available options
For Use With (Equipment):
Beckman Coulter Analyzer
This item is not returnable. View return policy

Catalog No. 21-280-089

Supplier: Beckman Coulter C67723

This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.

This item is not returnable. View return policy

The DxH 560 will help save time and resources, deliver high-quality, accurate results, and improve patient and physician satisfaction.

  • Small footprint
  • Low amount of reagents
  • Streamlined workflow that executes in any commend in 3 steps or less
  • Integrated user interface
TRUSTED_SUSTAINABILITY

Specifications

Type Autoloader
For Use With (Equipment) Beckman Coulter Analyzer
Throughput 55 to 60 Samples/hr.
Depth (English) 18.1 in.
Depth (Metric) 46 cm
Height (English) 17.3 in.
Height (Metric) 44 cm
Width (English) 19.7 in.
Width (Metric) 50 cm
Weight (English) 48.5 lb.
Weight (Metric) 22 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.