Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter CellaVision Digital Morphology DM9600

Catalog No. 21403583
Click to view available options
Throughput:
30 Slides/hr.

CellaVision Digital Morphology Solutions help increase the efficiency of hematology workflow systems for small and mid- to large-volume laboratories in hospital and healthcare networks.

Faster slide reviews

  • CellaVision Digital Morphology Solutions allow you to reduce turnaround times with automated pre-classification of the WBC differential, RBC morphology and platelet estimates.
  • Remote review expedites expert opinions, making more efficient use of experienced morphologists' time.

Standardized differential results

  • Automatic cell location and pre-classification reduce the subjectivity of cell identification and improve the quality of results.

Remote slide reviews

  • Cell images are stored in a centralized database where they can be accessed, reviewed and reported on from any laboratory or physician's office location within the network.

Integrated with LIS

  • Cell images can be stored with patient records, making it easy to track patient progress over time.

Enhance ergonomics

  • CellaVision Digital Morphology solutions analyzers do not cause neck, wrist or eye fatigue like microscopy does. As a result, you can help preserve your staff's health and improve their satisfaction.
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Blood
For Use With (Equipment) Beckman Coulter Analyzers
Throughput 30 Slides/hr.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.