Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beckman Coulter DxM 1096 MicroScan WalkAway ID/AST System
Shop All Beckman Coulter Products
Click to view available options
Includes:
LabPro Low Voltage
UPS
Description
- Reduced turnaround time for rapid and specialty ID panels.
- Proven solutions through scientific expertise and proprietary antibiotic formulations.
- Reduced offline testing provides faster turnaround time.
- Customizable data management provides greater flexibility more actionable results.
- Reduced labor and material costs through fewer confirmatory and repeat tests.
- Lower cost of ownership with wide range of susceptibility tests on one panel.
- Streamlined workflow supports testing for most routine samples through a more efficient Prompt™ system.
- Simultaneous processing of conventional, rapid and specialty panels on a single, automated platform.
- Easy-to-view external LED indicators show status at a glance.
- Simplified maintenance with updated reagent-handling system.

Specifications
Specifications
For Use With (Equipment) | Beckman Coulter Analyzers |
Includes | LabPro Low Voltage |
Depth (English) | 34 in. |
Height (English) | 37 in. |
Width (English) | 38.5 in. |
Weight (English) | 410 lb. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction