Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter DxM 1096 MicroScan WalkAway ID/AST System

Catalog No. p-200106330 Shop All Beckman Coulter Products
Click to view available options
Includes:
LabPro Low Voltage
UPS

Catalog No. 21-403-586

Supplier: Beckman Coulter C12701

Only null left
Add to Cart
Add to Cart
Request a Quote

Enable your laboratory to optimize patient care while reducing risks, costs and operational burden associated with detecting emerging antimicrobial resistance.

  • Reduced turnaround time for rapid and specialty ID panels.
  • Proven solutions through scientific expertise and proprietary antibiotic formulations.
  • Reduced offline testing provides faster turnaround time.
  • Customizable data management provides greater flexibility more actionable results.
  • Reduced labor and material costs through fewer confirmatory and repeat tests.
  • Lower cost of ownership with wide range of susceptibility tests on one panel.
  • Streamlined workflow supports testing for most routine samples through a more efficient Prompt™ system.
  • Simultaneous processing of conventional, rapid and specialty panels on a single, automated platform.
  • Easy-to-view external LED indicators show status at a glance.
  • Simplified maintenance with updated reagent-handling system.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Beckman Coulter Analyzers
Includes LabPro Low Voltage
Depth (English) 34 in.
Height (English) 37 in.
Width (English) 38.5 in.
Weight (English) 410 lb.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.