Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DXR Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144466
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144466 Supplier Invitrogen™ Supplier No. PA5144466
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DXR Polyclonal Antibody for Western Blot, ELISA

Catalyzes the NADPH-dependent rearrangement and reduction of 1-deoxy-D-xylulose-5-phosphate (DXP) to 2-C-methyl-D-erythritol 4-phosphate (MEP).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DXR
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene dxr
Gene Accession No. P45568
Gene Alias ispC; yaeM
Gene Symbols dxr
Host Species Rabbit
Immunogen Recombinant Escherichia coli (strain K12) DXR protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 75202014
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.