Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DYDC1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142537
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
This gene encodes a member of a family of proteins that contains a DPY30 domain. The encoded protein is involved in acrosome formation during spermatid development. This gene locus overlaps with a closely related gene on the opposite strand. Alternative splicing results in multiple transcript variants.
Specifications
| DYDC1 | |
| Polyclonal | |
| Unconjugated | |
| DYDC1 | |
| 1700029M23Rik; bA36D19.5; BC019250 protein; DPY30 domain containing 1; DPY30 domain-containing protein 1; DPY30D1; DYDC1; FLJ43920; OTTHUMP00000019963; RSD9; RSD-9 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 143241 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8WWB3 | |
| DYDC1 | |
| Peptide with sequence C-RNEDILHSEEATLD. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction