Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYDC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988434
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8434 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8434 Supplier Bioss Supplier No. BS8286R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of a family of proteins that contains a DPY30 domain. The encoded protein is involved in acrosome formation during spermatid development. This gene locus overlaps with a closely related gene on the opposite strand. Alternative splicing results in multiple transcript variants.

Specifications

Antigen DYDC1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DYDC1
Gene Accession No. Q8WWB3
Gene Alias 1700029M23Rik; bA36D19.5; BC019250 protein; DPY30 domain containing 1; DPY30 domain-containing protein 1; DPY30D1; Dydc1; FLJ43920; OTTHUMP00000019963; RSD9; RSD-9
Gene Symbols DYDC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DYDC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143241
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.