Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYDC2 Antibody (04), Alexa Fluor™ 532 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306212AF532
Description
DYDC2 Monoclonal antibody specifically detects DYDC2 in Human samples. It is validated for ELISASpecifications
DYDC2 | |
Monoclonal | |
Alexa Fluor 532 | |
bA36D19.6, DPY30 domain containing 2, DPY30 domain-containing protein 2, MGC16186 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human DYDC2 (Q96IM9; Thr102-Pro176). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
04 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
84332 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction