Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYDC2 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300214DL594
Description
DYDC2 Polyclonal specifically detects DYDC2 in Human samples. It is validated for ELISA.Specifications
DYDC2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
84332 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
DyLight 594 | |
bA36D19.6, DPY30 domain containing 2, DPY30 domain-containing protein 2, MGC16186 | |
Produced in rabbits immunized with purified, recombinant DYDC2 (Q96IM9; Thr102-Pro176). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction