Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYDC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558208 Supplier Invitrogen™ Supplier No. PA558208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAHWLYHYRK TAKAKEENRE KKIHLQEEYD SSLKEMEMTE MLKQEEYQIQ QNCEKCHKEL TSETVSTKKT Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 50%.

DYDC2 encodes a member of a family of proteins that contains a DPY30 domain. This gene locus overlaps with a closely related gene on the opposite strand. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYDC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DYDC2
Gene Accession No. Q96IM9
Gene Alias bA36D19.6; DPY30 domain containing 2; DPY30 domain-containing protein 2; DYDC2; MGC16186; SPS1; SRML1
Gene Symbols DYDC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DYDC2. Recombinant protein control fragment (Product #RP-102797).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84332
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.