Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYKDDDDK Epitope Tag Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600345AF700
Description
DYKDDDDK Epitope Tag Polyclonal antibody specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.Specifications
DYKDDDDK Epitope Tag | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag | |
To produce DYKDDDDK Epitope Tag Antibody, Rabbits were immunized with ECS enterokinase cleavage site (xxxDDDDK) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. | |
Affinity Purified | |
RUO | |
DYKDDDDK Epitope Tag Antibody reacts with FLAG. | |
IgG |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rabbit | |
1.01 kDa | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
Epitope Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction