Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYKDDDDK Epitope Tag Horseradish Peroxidase-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. HAM85291
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HAM85291 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. HAM85291 Supplier R&D Systems Supplier No. HAM85291
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DYKDDDDK Epitope Tag Monoclonal specifically detects DYKDDDDK Epitope Tag in Multi-Species samples. It is validated for Western Blot.

Specifications

Antigen DYKDDDDK Epitope Tag
Applications Western Blot
Classification Monoclonal
Clone 1042E
Conjugate HRP
Dilution Western Blot 1:1000-1:3000 dilution
Formulation Supplied as a 0.2 μm filtered solution in PBS with No Preservative
Gene Alias DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag
Host Species Rabbit
Immunogen DYKDDDDK epitope tag
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects N-terminal and C-terminal DYKDDDDK (FLAG™) epitope tagged proteins in Western blot.
Target Species Multi-species
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.