Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYKDDDDK Epitope Tag Antibody (L5), HRP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP106712H0.1ML
Description
DYKDDDDK Epitope Tag Monoclonal specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
DYKDDDDK Epitope Tag | |
Monoclonal | |
HRP | |
PBS with no preservative | |
Rat | |
0.1 mL | |
Human, Drosophila, Guinea Pig, Non-species specific | |
IgG2a |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
L5 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
DDDDK epitope tag, DDK, DYKDDDDK epitope tag, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag | |
N-terminal DYKDDDDK-tagged extracellular domain of mouse Langerin. Binds to same epitope as Sigma′s Anti-FLAG&No.174; M2 Antibody. | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction