Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYKDDDDK Tag Monoclonal Antibody (5A8E5), THE™, GenScript™

Catalog No. 50272476
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-272-476 200 μg
50-272-474 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-272-476 Supplier Genscript Corporation Supplier No. A00187200
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/mL. The DYKDDDDK peptide is a small component of the epitope which does not appear to interfere with the bioactivity or the biodistribution of the recombinant protein. It has been used extensively as a general epitope tag in expression vectors. THE™ DYKDDDDK Tag Antibody, mAb, Mouse is a high-affinity monoclonal antibody that can be used to detect DYKDDDDK-tagged proteins.

DYKDDDDK Tag Monoclonal antibody specifically detects DYKDDDDK Tag in Tag samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen DYKDDDDK Tag
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5A8E5
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag
Host Species Mouse
Immunogen A synthetic peptide (DYKDDDDK) coupled to KLH
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG2b κ
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.